Gene Transcripts

Gene Symbol
NAB1
Entrez Gene ID
4664
Gene Name
NGFI-A binding protein 1
Gene Aliases
Chromosome Location
Chr. 2: 190648860 - 190692766 on Build GRCh38
UniGene ID
Hs.107474
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321312.1 NP_001308241.1 2 418
NM_001321313.1 NP_001308242.1 2 204
NM_001321314.1 NP_001308243.1 1 127
NM_001321315.1 NP_001308244.1 1 127
NM_005966.3 NP_005957.2 4 680
XM_005246582.1 XP_005246639.1 3 353
XM_005246583.1 XP_005246640.1 3 467
XM_005246586.1 XP_005246643.1 2 160
XM_005246587.1 XP_005246644.1 2 160
XM_011511219.2 XP_011509521.1 3 322
XM_017004170.1 XP_016859659.1 3 363
XM_017004171.1 XP_016859660.1 2 533
XM_017004172.1 XP_016859661.1 2 305
XM_017004173.1 XP_016859662.1 3 388
XM_017004174.1 XP_016859663.1 2 204
XM_017004175.1 XP_016859664.1 2 369
XM_017004176.1 XP_016859665.1 2 272
GenBank mRNA AF045451.1 - 1 127
AF045452.1 - 1 127
AK292446.1 - 2 218
AK313542.1 - 1 225
BC035724.1 - 4 569 5721972
DB457461.1 - 2 418
U47007.1 - 1 127

View Full Product Details