Gene Transcripts
- Gene Symbol
- TMOD1
- Entrez Gene ID
- 7111
- Gene Name
- tropomodulin 1
- Gene Aliases
- D9S57E, ETMOD, TMOD
- Chromosome Location
- Chr. 9: 97501180 - 97601743 on Build GRCh38
- UniGene ID
- Hs.404289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BU783086.1 |
- |
1 |
28 |
|
- Gene Symbol
- TSTD2
- Entrez Gene ID
- 158427
- Gene Name
- thiosulfate sulfurtransferase like domain containing 2
- Gene Aliases
- C9orf97
- Chromosome Location
- Chr. 9: 97600080 - 97633680 on Build GRCh38
- UniGene ID
- Hs.723236
- Species
- Homo sapiens
View Full Product Details