Gene Transcripts

Gene Symbol
GLT1D1
Entrez Gene ID
144423
Gene Name
glycosyltransferase 1 domain containing 1
Gene Aliases
Chromosome Location
Chr. 12: 128853323 - 128984964 on Build GRCh38
UniGene ID
Hs.655668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144669.2 NP_653270.1 2 264
NR_133646.1 2 264
XM_005253553.4 XP_005253610.2 2 375
XM_011537956.2 XP_011536258.2 2 375
XM_017018855.1 XP_016874344.1 2 375
XM_017018857.1 XP_016874346.1 2 131
XM_017018858.1 XP_016874347.1 2 131
XM_017018859.1 XP_016874348.1 2 375
XM_017018860.1 XP_016874349.1 2 131
XM_017018861.1 XP_016874350.1 2 163
GenBank mRNA AK056540.1 - 2 162
AK295333.1 - 2 92
BC043528.1 - 2 264 5168267

View Full Product Details