Gene Transcripts

Gene Symbol
AXDND1
Entrez Gene ID
126859
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
C1orf125
Chromosome Location
Chr. 1: 179365705 - 179554735 on Build GRCh38
UniGene ID
Hs.658505
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144696.5 NP_653297.3 8 1115
NR_073544.1 8 989
XM_011509165.1 XP_011507467.1 8 1094
XM_011509166.2 XP_011507468.1 8 1141
XM_011509167.2 XP_011507469.1 8 1256
XM_011509168.1 XP_011507470.1 8 1098
XM_011509169.1 XP_011507471.1 7 887
XM_011509170.1 XP_011507472.1 8 1094
XM_011509171.1 XP_011507473.1 7 912
XM_011509174.1 XP_011507476.1 8 1094
XM_011509175.1 XP_011507477.1 8 1094
XM_011509176.1 XP_011507478.1 8 1094
XM_011509178.2 XP_011507480.1 8 1094
XM_011509179.2 XP_011507481.1 3 202
XM_011509180.1 XP_011507482.1 8 1094
GenBank mRNA AK057502.1 - 6 482
AK302972.1 - 8 1094
AL833914.1 - 6 542
BC146823.1 - 8 990 8860438
BX647935.1 - 8 1116

View Full Product Details