Gene Transcripts
- Gene Symbol
- RBM11
- Entrez Gene ID
- 54033
- Gene Name
- RNA binding motif protein 11
- Gene Aliases
- Chromosome Location
- Chr. 21: 14216107 - 14228372 on Build GRCh38
- UniGene ID
- Hs.283828
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001320602.1 |
NP_001307531.1
|
3, 4 |
397 |
|
NM_144770.4 |
NP_658983.3
|
3, 4 |
397 |
|
NR_135308.1 |
|
2, 3 |
234 |
|
NR_135309.1 |
|
2, 3 |
234 |
|
NR_135310.1 |
|
3, 4 |
407 |
|
NR_135311.1 |
|
3, 4 |
407 |
|
XM_005260997.4 |
XP_005261054.1
|
3, 4 |
420 |
|
XM_017028386.1 |
XP_016883875.1
|
3, 4 |
552 |
|
XM_017028387.1 |
XP_016883876.1
|
3, 4 |
559 |
|
GenBank mRNA |
AK091331.1 |
- |
3, 4 |
379 |
|
BC030196.1 |
- |
3, 4 |
349 |
5199053 |
View Full Product Details