Gene Transcripts

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
Chromosome Location
Chr. 11: 34150987 - 34358008 on Build GRCh38
UniGene ID
Hs.23361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145804.2 NP_665803.2 8, 9 2285
GenBank mRNA AK091560.1 - 5, 6 503
AK095632.1 - 8, 9 1532
AK291744.1 - 8, 9 2286
BC093968.1 - 8, 9 1397 7939813

View Full Product Details