Gene Transcripts

Gene Symbol
PIDD1
Entrez Gene ID
55367
Gene Name
p53-induced death domain protein 1
Gene Aliases
LRDD, PIDD
Chromosome Location
Chr. 11: 799179 - 809504 on Build GRCh38
UniGene ID
Hs.592290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145886.3 NP_665893.2 2 344
NM_145887.3 NP_665894.2 2 344
XM_005253005.4 XP_005253062.1 3 1282
XM_005253006.4 XP_005253063.1 3 845
XM_005253007.3 XP_005253064.1 2 362
XM_005253008.4 XP_005253065.1 3 1282
XM_011520209.2 XP_011518511.1 2 580
XM_011520210.2 XP_011518512.1 3 1282
XM_011520211.2 XP_011518513.1 3 1282
XM_011520212.1 XP_011518514.1 2 362
GenBank mRNA AB208832.1 - 2 360
AB208949.1 - 2 324
AB209529.1 - 1 912
AF229178.1 - 1 486
AF274972.1 - 2 340
BC014904.1 - 1 282 3908102
BG723687.1 - 2 406
BQ059356.1 - 2 331
CN427944.1 - 1 278
CN427946.1 - 2 343

View Full Product Details