Gene Transcripts

Gene Symbol
NXNL2
Entrez Gene ID
158046
Gene Name
nucleoredoxin-like 2
Gene Aliases
C9orf121, RDCVF2
Chromosome Location
Chr. 9: 88535088 - 88584510 on Build GRCh38
UniGene ID
Hs.734507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161625.1 NP_001155097.1 1 573
NM_145283.2 NP_660326.2 1 573
XM_005251727.3 XP_005251784.1 1 579
XM_011518275.2 XP_011516577.1 1 576
XM_011518276.2 XP_011516578.1 1 586
GenBank mRNA BC022521.1 - 1 569
BI517442.1 - 1 481

View Full Product Details