Gene Transcripts

Gene Symbol
PIK3AP1
Entrez Gene ID
118788
Gene Name
phosphoinositide-3-kinase adaptor protein 1
Gene Aliases
BCAP
Chromosome Location
Chr. 10: 96593312 - 96720522 on Build GRCh38
UniGene ID
Hs.310456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152309.2 NP_689522.2 17 2803
XM_005269498.1 XP_005269555.1 16 2326
XM_005269499.1 XP_005269556.1 16 2350
XM_011539248.1 XP_011537550.1 16 2676
XM_011539249.1 XP_011537551.1 16 2639
GenBank mRNA AK092883.1 - 16 2326
AK122584.1 - 6 3632
AK125635.1 - 17 2795
BC029917.1 - 10 1654 5168493
BX648550.1 - 17 2699

View Full Product Details