Gene Transcripts

Gene Symbol
FGFBP3
Entrez Gene ID
143282
Gene Name
fibroblast growth factor binding protein 3
Gene Aliases
C10orf13, FGF-BP3
Chromosome Location
Chr. 10: 91906588 - 91909501 on Build GRCh38
UniGene ID
Hs.466120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152429.4 NP_689642.3 2 1105
GenBank mRNA AK075410.1 - 2 1088
BC025966.1 - 2 1074 4895171

View Full Product Details