Gene Transcripts

Gene Symbol
PDE6B
Entrez Gene ID
5158
Gene Name
phosphodiesterase 6B
Gene Aliases
CSNB3, CSNBAD2, PDEB, RP40, rd1
Chromosome Location
Chr. 4: 587593 - 670892 on Build GRCh38
UniGene ID
Hs.623810
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000283.3 NP_000274.2 1 253
NM_001145291.1 NP_001138763.1 1 253
XM_011513473.2 XP_011511775.1 2 1520
XM_011513474.2 XP_011511776.1 2 1520
XM_011513475.2 XP_011511777.1 4 1623
XM_011513476.2 XP_011511778.1 2 1520
GenBank mRNA BC000249.1 - 1 243
BT009794.1 - 1 200
S41458.1 - 1 221
X66142.1 - 1 205

View Full Product Details