Gene Transcripts

Gene Symbol
SNX31
Entrez Gene ID
169166
Gene Name
sorting nexin 31
Gene Aliases
Chromosome Location
Chr. 8: 100572882 - 100663410 on Build GRCh38
UniGene ID
Hs.567758
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152628.3 NP_689841.3 14 2274
XM_005250815.3 XP_005250872.1 11 1913
XM_011516899.1 XP_011515201.1 13 2251
XM_017013153.1 XP_016868642.1 14 2271
XM_017013154.1 XP_016868643.1 14 2271
XM_017013155.1 XP_016868644.1 14 2268
XM_017013156.1 XP_016868645.1 13 2217
XM_017013157.1 XP_016868646.1 13 2160
XM_017013158.1 XP_016868647.1 13 2157
XM_017013159.1 XP_016868648.1 12 2103
XM_017013160.1 XP_016868649.1 13 2070
XM_017013161.1 XP_016868650.1 11 2025
XM_017013162.1 XP_016868651.1 12 1956
XM_017013163.1 XP_016868652.1 11 2354
GenBank mRNA AK093906.1 - 14 2274

View Full Product Details