Gene Transcripts

Gene Symbol
BEND6
Entrez Gene ID
221336
Gene Name
BEN domain containing 6
Gene Aliases
C6orf65
Chromosome Location
Chr. 6: 56954975 - 57027652 on Build GRCh38
UniGene ID
Hs.582993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318539.1 NP_001305468.1 2, 3 709
NM_152731.2 NP_689944.2 2, 3 709
XM_011514346.2 XP_011512648.1 2, 3 697
XM_017010406.1 XP_016865895.1 2, 3 260
XM_017010407.1 XP_016865896.1 2, 3 243
GenBank mRNA AK054724.1 - 2, 3 394
AK303705.1 - 1, 2 79
BC036119.1 - 2, 3 558
DC411761.1 - 1, 2 79

View Full Product Details