Gene Transcripts

Gene Symbol
CASP6
Entrez Gene ID
839
Gene Name
caspase 6
Gene Aliases
MCH2
Chromosome Location
Chr. 4: 109688629 - 109703473 on Build GRCh38
UniGene ID
Hs.654616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001226.3 NP_001217.2 5 519
NM_032992.2 NP_116787.1 2 252
NR_133012.1 4 442
GenBank mRNA BC000305.2 - 5 467
BC004460.1 - 5 467 2820468
BM837197.1 - 2 198
BX380795.2 - 4 383
U20536.1 - 5 519
U20537.1 - 2 174

View Full Product Details