Gene Transcripts

Gene Symbol
CRYBG3
Entrez Gene ID
131544
Gene Name
crystallin beta-gamma domain containing 3
Gene Aliases
DKFZp667G2110, vlAKAP
Chromosome Location
Chr. 3: 97821930 - 97944986 on Build GRCh38
UniGene ID
Hs.733378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153605.3 NP_705833.3 22 9692
XM_005247117.4 XP_005247174.1 21 9395
GenBank mRNA AF451988.1 - 10 1809
AK091199.1 - 14 2163
AL831927.1 - 10 1718
BC036247.1 - 5 1237
BC050394.1 - 7 1475
BX647079.1 - 14 2183
BX648353.1 - 19 3745
CR749313.1 - 19 2820

View Full Product Details