Gene Transcripts

Gene Symbol
KREMEN2
Entrez Gene ID
79412
Gene Name
kringle containing transmembrane protein 2
Gene Aliases
KRM2
Chromosome Location
Chr. 16: 2964216 - 2968383 on Build GRCh38
UniGene ID
Hs.661128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253725.1 NP_001240654.1 8 1808
NM_001253726.1 NP_001240655.1 9 1887
NM_024507.3 NP_078783.1 8 1925
NM_172229.2 NP_757384.1 9 2004
GenBank mRNA AK027669.1 - 8 1829
AK075033.1 - 8 1749
AK223422.1 - 8 1697
AK301953.1 - 9 1828
BC003533.1 - 8 1732 3609280
BC009383.2 - 7 2289 4127918

View Full Product Details