Gene Transcripts

Gene Symbol
C18orf54
Entrez Gene ID
162681
Gene Name
chromosome 18 open reading frame 54
Gene Aliases
LAS2
Chromosome Location
Chr. 18: 54357914 - 54382035 on Build GRCh38
UniGene ID
Hs.208701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288980.1 NP_001275909.1 8, 9 1840
NM_001288981.1 NP_001275910.1 8, 9 1672
NM_001288982.1 NP_001275911.1 6, 7 1043
NM_173529.5 NP_775800.3 7, 8 1198
NR_110234.1 5, 6 524
XM_005258201.1 XP_005258258.1 7, 8 2541
XM_005258203.1 XP_005258260.1 7, 8 2001
XM_017025567.1 XP_016881056.1 7, 8 2224
GenBank mRNA AB267906.1 - 7, 8 2536
AB330143.1 - 7, 8 2053
AB330144.1 - 7, 8 1996
AB353076.1 - 8, 9 1839
AB353077.1 - 8, 9 1671
AK126503.1 - 6, 7 1278
BC036054.2 - 6, 7 1100 5273569
BX640877.1 - 7, 8 1171
CN359540.1 - 5, 6 528

View Full Product Details