Gene Transcripts

Gene Symbol
UNC5CL
Entrez Gene ID
222643
Gene Name
unc-5 family C-terminal like
Gene Aliases
MUXA, ZUD
Chromosome Location
Chr. 6: 41026901 - 41039199 on Build GRCh38
UniGene ID
Hs.158357
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173561.2 NP_775832.2 9 2752
GenBank mRNA BC035284.1 - 9 2742
JF681947.1 - 9 2770

View Full Product Details