Gene Transcripts

Gene Symbol
RSPO1
Entrez Gene ID
284654
Gene Name
R-spondin 1
Gene Aliases
CRISTIN3, RSPO
Chromosome Location
Chr. 1: 37611350 - 37634923 on Build GRCh38
UniGene ID
Hs.135015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001038633.3 NP_001033722.1 8 1712
NM_001242908.1 NP_001229837.1 7 1533
NM_001242909.1 NP_001229838.1 7 1212
NM_001242910.1 NP_001229839.1 6 1344
XM_006710583.3 XP_006710646.1 6 1448
GenBank mRNA AK098225.1 - 7 954
BC114966.1 - 6 1130 40084277
DQ165084.1 - 7 1275
DQ165085.1 - 6 1086
DQ318235.1 - 8 1422

View Full Product Details