Gene Transcripts

Gene Symbol
C8orf31
Entrez Gene ID
286122
Gene Name
chromosome 8 open reading frame 31
Gene Aliases
Chromosome Location
Chr. 8: 143038983 - 143054303 on Build GRCh38
UniGene ID
Hs.660382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173687.3 NP_775958.1 6 1215
NR_126398.1 8 1397
NR_126399.1 6 1206
GenBank mRNA AK094450.1 - 6 1163
BC120993.2 - 6 1141

View Full Product Details