Gene Transcripts

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
Chromosome Location
Chr. 12: 50504985 - 50748667 on Build GRCh38
UniGene ID
Hs.505516
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173602.2 NP_775873.2 7 1043
XM_005269044.1 XP_005269101.1 7 1043
XM_006719520.3 XP_006719583.1 8 1019
XM_006719521.3 XP_006719584.1 7 856
XM_011538594.2 XP_011536896.1 8 1069
XM_011538595.2 XP_011536897.1 8 1069
XM_011538596.2 XP_011536898.1 7 856
XM_017019699.1 XP_016875188.1 8 1069
XM_017019700.1 XP_016875189.1 7 1043
XM_017019701.1 XP_016875190.1 7 1043
XM_017019702.1 XP_016875191.1 7 1043
XM_017019703.1 XP_016875192.1 6 819
XM_017019704.1 XP_016875193.1 6 830
XM_017019705.1 XP_016875194.1 6 830
GenBank mRNA AK091597.1 - 7 1043
BC075027.2 - 7 921
BX537905.1 - 6 664

View Full Product Details