Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 1834 | |
NM_001160258.1 | NP_001153730.1 | 4 | 1872 | ||
NM_001160259.1 | NP_001153731.1 | 2 | 1747 | ||
NM_016083.4 | NP_057167.2 | 2 | 1691 | ||
NM_033181.3 | NP_149421.2 | 2 | 1346 | ||
XM_006715330.3 | XP_006715393.1 | 2 | 1638 | ||
XM_011535424.1 | XP_011533726.1 | 3 | 2178 | ||
XM_011535425.1 | XP_011533727.1 | 4 | 2216 | ||
XM_011535426.1 | XP_011533728.1 | 5 | 2374 | ||
XM_011535428.1 | XP_011533730.1 | 3 | 2025 | ||
XM_017010238.1 | XP_016865727.1 | 4 | 5002 | ||
XM_017010239.1 | XP_016865728.1 | 3 | 3325 | ||
XM_017010240.1 | XP_016865729.1 | 2 | 1685 | ||
GenBank mRNA | AF107262.1 | - | 1 | 1391 | |
AK313908.1 | - | 2 | 1512 | ||
AY766182.1 | - | 2 | 1283 | ||
AY995204.1 | - | 1 | 1382 | ||
BC074811.2 | - | 1 | 1420 | 30915316 | |
BC074812.2 | - | 1 | 1420 | 30915532 | |
BC095513.1 | - | 1 | 1379 | ||
BC100968.1 | - | 1 | 1382 | 40004970 | |
BC100969.1 | - | 1 | 1382 | 40004975 | |
BC100970.1 | - | 1 | 1382 | 40004978 | |
BC100971.1 | - | 1 | 1382 | 40004979 | |
DQ067455.1 | - | 1 | 1382 | ||
KC292264.1 | - | 2 | 1346 | ||
X54937.1 | - | 2 | 1530 | ||
X81120.1 | - | 1 | 1473 | ||
X81121.1 | - | 2 | 1215 |