Gene Transcripts
- Gene Symbol
- SPEG
- Entrez Gene ID
- 10290
- Gene Name
- SPEG complex locus
- Gene Aliases
- APEG-1, APEG1, BPEG, CNM5, SPEGalpha, SPEGbeta
- Chromosome Location
- Chr. 2: 219434215 - 219493632 on Build GRCh38
- UniGene ID
- Hs.21639
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001173476.1 |
NP_001166947.1
|
2, 3 |
369 |
|
NM_005876.4 |
NP_005867.3
|
7, 8 |
2624 |
|
XM_005246237.2 |
XP_005246294.1
|
9, 10 |
2352 |
|
XM_005246239.2 |
XP_005246296.1
|
9, 10 |
2167 |
|
XM_005246240.2 |
XP_005246297.1
|
2, 3 |
369 |
|
XM_005246241.1 |
XP_005246298.1
|
2, 3 |
263 |
|
XM_005246242.4 |
XP_005246299.1
|
2, 3 |
388 |
|
XM_006712189.3 |
XP_006712252.1
|
8, 9 |
2357 |
|
XM_006712193.3 |
XP_006712256.1
|
2, 3 |
3488 |
|
XM_011510479.2 |
XP_011508781.1
|
8, 9 |
2790 |
|
XM_011510483.2 |
XP_011508785.2
|
8, 9 |
2387 |
|
XM_017003157.1 |
XP_016858646.1
|
8, 9 |
2387 |
|
XM_017003158.1 |
XP_016858647.1
|
4, 5 |
1963 |
|
XM_017003159.1 |
XP_016858648.1
|
5, 6 |
2146 |
|
XM_017003161.1 |
XP_016858650.1
|
5, 6 |
2146 |
|
XM_017003162.1 |
XP_016858651.1
|
5, 6 |
2146 |
|
GenBank mRNA |
AK055387.1 |
- |
3, 4 |
977 |
|
AK126500.1 |
- |
8, 9 |
2812 |
|
AK128689.1 |
- |
7, 8 |
2622 |
|
AK129604.1 |
- |
1, 2 |
826 |
|
AK289531.1 |
- |
9, 10 |
2149 |
|
AY603755.1 |
- |
6, 7 |
2237 |
|
BC006346.2 |
- |
2, 3 |
211 |
|
CR542201.1 |
- |
1, 2 |
77 |
|
DA063220.1 |
- |
2, 3 |
370 |
|
U57099.1 |
- |
2, 3 |
202 |
|
View Full Product Details