Gene Transcripts

Gene Symbol
MLKL
Entrez Gene ID
197259
Gene Name
mixed lineage kinase domain-like
Gene Aliases
hMLKL
Chromosome Location
Chr. 16: 74671855 - 74701187 on Build GRCh38
UniGene ID
Hs.119878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152649.3 NP_689862.1 3, 4 1240
XM_005255834.3 XP_005255891.1 4, 5 1139
XM_011522936.2 XP_011521238.1 3, 4 1279
GenBank mRNA AK091708.1 - 3, 4 983

View Full Product Details