Gene Transcripts

Gene Symbol
SFT2D1
Entrez Gene ID
113402
Gene Name
SFT2 domain containing 1
Gene Aliases
C6orf83, pRGR1
Chromosome Location
Chr. 6: 166319728 - 166342606 on Build GRCh38
UniGene ID
Hs.487143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145169.2 NP_660152.1 3, 4 366
NR_130112.1 4, 5 389
NR_130113.1 4, 5 389
NR_130114.1 4, 5 380
GenBank mRNA AF041429.1 - 3, 4 300
AW964442.1 - 4, 5 311
BC018969.1 - 3, 4 271 3959886
BC028388.1 - 4, 5 333
BF223532.1 - 4, 5 284
BG706927.1 - 3, 4 355
BI597156.1 - 4, 5 351
DB476616.1 - 4, 5 389

View Full Product Details