Gene Transcripts

Gene Symbol
PAQR3
Entrez Gene ID
152559
Gene Name
progestin and adipoQ receptor family member 3
Gene Aliases
RKTG
Chromosome Location
Chr. 4: 78887225 - 78939428 on Build GRCh38
UniGene ID
Hs.657312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005262769.4 XP_005262826.1 5, 6 992
XM_017007785.1 XP_016863274.1 5, 6 992
GenBank mRNA BC047510.1 - 5, 6 967

View Full Product Details