Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002268.4 | NP_002259.1 | 2 | 394 | |
GenBank mRNA | AB002533.2 | - | 2 | 97 | |
AK291041.1 | - | 2 | 346 | ||
BC028691.2 | - | 2 | 339 | 4838709 | |
BC034493.2 | - | 2 | 394 | 4821602 | |
U93240.1 | - | 2 | 238 |