Gene Transcripts

Gene Symbol
PRRC2C
Entrez Gene ID
23215
Gene Name
proline rich coiled-coil 2C
Gene Aliases
BAT2-iso, BAT2D1, BAT2L2, XTP2
Chromosome Location
Chr. 1: 171485496 - 171593511 on Build GRCh38
UniGene ID
Hs.494614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015172.3 NP_055987.2 2, 3 376
XM_005245016.4 XP_005245073.1 2, 3 393
XM_006711238.3 XP_006711301.1 2, 3 393
XM_017000760.1 XP_016856249.1 2, 3 393
GenBank mRNA AF488829.1 - 1, 2 110
AL096857.1 - 1, 2 167
BC001260.1 - 2, 3 347

View Full Product Details