Gene Transcripts

Gene Symbol
SREK1IP1
Entrez Gene ID
285672
Gene Name
SREK1 interacting protein 1
Gene Aliases
P18SRP, SFRS12IP1
Chromosome Location
Chr. 5: 64718148 - 64768849 on Build GRCh38
UniGene ID
Hs.69504
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173829.3 NP_776190.1 3, 4 349
XM_011543344.2 XP_011541646.1 2, 3 481
GenBank mRNA AK094073.1 - 349
AL832348.1 - 3, 4 306
BC108716.1 - 3, 4 279 6184559
BC127085.1 - 3, 4 287 40122236
BC127086.1 - 3, 4 287 40122239

View Full Product Details