Gene Transcripts

Gene Symbol
IER3IP1
Entrez Gene ID
51124
Gene Name
immediate early response 3 interacting protein 1
Gene Aliases
HSPC039, MEDS, PRO2309
Chromosome Location
Chr. 18: 47155019 - 47176374 on Build GRCh38
UniGene ID
Hs.130917
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016097.4 NP_057181.1 2, 3 298
GenBank mRNA AF125100.1 - 2, 3 281
AF164798.1 - 2, 3 267
AF371963.1 - 2, 3 271
AK027108.1 - 2, 3 288
AL136667.1 - 2, 3 278
BC010888.1 - 2, 3 264 4285974
BC017391.1 - 2, 3 231 4133818
BM829309.1 - 2, 3 297
CR533488.1 - 2, 3 201

View Full Product Details