Gene Transcripts

Gene Symbol
HOPX
Entrez Gene ID
84525
Gene Name
HOP homeobox
Gene Aliases
CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO
Chromosome Location
Chr. 4: 56647988 - 56681866 on Build GRCh38
UniGene ID
Hs.619396
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145459.1 NP_001138931.1 2, 3 378
NM_032495.5 NP_115884.4 3, 4 732
NM_139211.4 NP_631957.1 2, 3 334
NM_139212.3 NP_631958.1 2, 3 607
XM_017008728.1 XP_016864217.1 2, 3 369
XM_017008729.1 XP_016864218.1 2, 3 251
XM_017008730.1 XP_016864219.1 2, 3 299
XM_017008731.1 XP_016864220.1 2, 3 334
XM_017008732.1 XP_016864221.1 3, 4 610
XM_017008733.1 XP_016864222.1 2, 3 174
XM_017008734.1 XP_016864223.1 2, 3 230
GenBank mRNA AB019573.1 - 1, 2 44
AB059408.1 - 2, 3 292
AB059409.1 - 2, 3 175
AB059410.2 - 2, 3 184
AF454763.1 - 2, 3 197
AF492675.1 - 2, 3 581
AF492676.1 - 3, 4 313
AF492677.1 - 3, 4 357
AF492680.1 - 2, 3 333
AF492681.1 - 2, 3 296
AK289707.1 - 2, 3 276
AW955211.1 - 2, 3 300
BC014225.2 - 1, 2 447 4335211
CB996541.1 - 2, 3 370
EB386856.1 - 2, 3 328

View Full Product Details