Gene Transcripts

Gene Symbol
LRRC1
Entrez Gene ID
55227
Gene Name
leucine rich repeat containing 1
Gene Aliases
LANO, dJ523E19.1
Chromosome Location
Chr. 6: 53794497 - 53924125 on Build GRCh38
UniGene ID
Hs.606493
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018214.4 NP_060684.4 1, 2 444
XM_011514726.2 XP_011513028.1 2, 3 808
XM_017010997.1 XP_016866486.1 1, 2 736
GenBank mRNA AF332199.1 - 1, 2 391
AF359380.1 - 1, 2 167
AK001637.1 - 1, 2 148
AK021896.1 - 2, 3 808
AK292248.1 - 1, 2 289
BC003193.1 - 1, 2 444 3504090

View Full Product Details