Gene Transcripts

Gene Symbol
LYPD6B
Entrez Gene ID
130576
Gene Name
LY6/PLAUR domain containing 6B
Gene Aliases
CT116, LYPD7
Chromosome Location
Chr. 2: 149038467 - 149215262 on Build GRCh38
UniGene ID
Hs.357567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317002.1 NP_001303931.1 5 383
NM_001317003.1 NP_001303932.1 3 493
NM_001317004.1 NP_001303933.1 4 294
NM_001317005.1 NP_001303934.1 4 554
NM_001317006.1 NP_001303935.1 2 422
NM_177964.4 NP_808879.2 4 565
XM_005246306.4 XP_005246363.1 6 535
XM_006712279.2 XP_006712342.1 4 387
XM_006712280.3 XP_006712343.1 6 533
XM_006712281.3 XP_006712344.1 4 307
XM_011510620.2 XP_011508922.1 6 408
XM_011510623.2 XP_011508925.1 5 381
XM_011510625.1 XP_011508927.1 7 612
XM_017003364.1 XP_016858853.1 5 393
XM_017003365.1 XP_016858854.1 4 320
XM_017003366.1 XP_016858855.1 4 364
GenBank mRNA AF435957.1 - 4 294
BC018203.1 - 3 242 3861272
BC040176.2 - 4 565 4794338
BQ056975.1 - 2 159
CN359800.1 - 4 254
DB461489.1 - 5 376

View Full Product Details