Gene Transcripts

Gene Symbol
TMTC1
Entrez Gene ID
83857
Gene Name
transmembrane and tetratricopeptide repeat containing 1
Gene Aliases
ARG99, OLF, TMTC1A
Chromosome Location
Chr. 12: 29500813 - 29784759 on Build GRCh38
UniGene ID
Hs.401954
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193451.1 NP_001180380.1 7 1213
NM_175861.3 NP_787057.2 7 1304
XM_005253499.4 XP_005253556.1 9 1465
XM_017020003.1 XP_016875492.1 9 1466
XM_017020004.1 XP_016875493.1 8 1258
XM_017020005.1 XP_016875494.1 6 1071
XM_017020006.1 XP_016875495.1 9 1469
XM_017020007.1 XP_016875496.1 5 491
GenBank mRNA AK055962.1 - 5 465
AK123619.1 - 3 895
AK127297.1 - 9 1275
BC042083.1 - 7 1304 5749757
GQ865601.1 - 7 1213

View Full Product Details