Gene Transcripts

Gene Symbol
CEBPD
Entrez Gene ID
1052
Gene Name
CCAAT/enhancer binding protein delta
Gene Aliases
C/EBP-delta, CELF, CRP3, NF-IL6-beta
Chromosome Location
Chr. 8: 47736914 - 47738164 on Build GRCh38
UniGene ID
Hs.440829
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005195.3 NP_005186.2 1 706
GenBank mRNA BC094715.1 - 2 918
BC105109.1 - 1 701 8144112
M83667.1 - 1 750

View Full Product Details