Gene Transcripts

Gene Symbol
ANP32A
Entrez Gene ID
8125
Gene Name
acidic nuclear phosphoprotein 32 family member A
Gene Aliases
C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32
Chromosome Location
Chr. 15: 68778535 - 68820922 on Build GRCh38
UniGene ID
Hs.458747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006305.3 NP_006296.1 2 322
GenBank mRNA AF025684.1 - 2 299
AK127498.1 - 2 3107
AK223280.1 - 2 268
AK312703.1 - 2 295
AY349171.1 - 2 151
BC000608.2 - 2 286
BC007200.1 - 2 284 3677623
BC106719.2 - 2 151 40014106
BC125143.1 - 2 155 40119966
BT007436.1 - 2 151
DA825897.1 - 2 322
U60823.1 - 2 151
U73477.1 - 2 247
X75090.1 - 2 254

View Full Product Details