Gene Transcripts
- Gene Symbol
- HOPX
- Entrez Gene ID
- 84525
- Gene Name
- HOP homeobox
- Gene Aliases
- CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO
- Chromosome Location
- Chr. 4: 56647988 - 56681866 on Build GRCh38
- UniGene ID
- Hs.619396
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145459.1 |
NP_001138931.1
|
2, 3 |
381 |
|
NM_032495.5 |
NP_115884.4
|
3, 4 |
735 |
|
NM_139211.4 |
NP_631957.1
|
2, 3 |
337 |
|
NM_139212.3 |
NP_631958.1
|
2, 3 |
610 |
|
XM_017008728.1 |
XP_016864217.1
|
2, 3 |
372 |
|
XM_017008729.1 |
XP_016864218.1
|
2, 3 |
254 |
|
XM_017008730.1 |
XP_016864219.1
|
2, 3 |
302 |
|
XM_017008731.1 |
XP_016864220.1
|
2, 3 |
337 |
|
XM_017008732.1 |
XP_016864221.1
|
3, 4 |
613 |
|
XM_017008733.1 |
XP_016864222.1
|
2, 3 |
177 |
|
XM_017008734.1 |
XP_016864223.1
|
2, 3 |
233 |
|
GenBank mRNA |
AB019573.1 |
- |
1, 2 |
47 |
|
AB059408.1 |
- |
2, 3 |
295 |
|
AB059409.1 |
- |
2, 3 |
178 |
|
AB059410.2 |
- |
2, 3 |
187 |
|
AF454763.1 |
- |
2, 3 |
200 |
|
AF492675.1 |
- |
2, 3 |
584 |
|
AF492676.1 |
- |
3, 4 |
316 |
|
AF492677.1 |
- |
3, 4 |
360 |
|
AF492680.1 |
- |
2, 3 |
336 |
|
AF492681.1 |
- |
2, 3 |
299 |
|
AK289707.1 |
- |
2, 3 |
279 |
|
AW955211.1 |
- |
2, 3 |
303 |
|
BC014225.2 |
- |
1, 2 |
450 |
4335211 |
CB996541.1 |
- |
2, 3 |
373 |
|
EB386856.1 |
- |
2, 3 |
331 |
|
View Full Product Details