Gene Transcripts
- Gene Symbol
- HOPX
- Entrez Gene ID
- 84525
- Gene Name
- HOP homeobox
- Gene Aliases
- CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO
- Chromosome Location
- Chr. 4: 56647988 - 56681866 on Build GRCh38
- UniGene ID
- Hs.619396
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145459.1 |
NP_001138931.1
|
2, 3 |
387 |
|
NM_032495.5 |
NP_115884.4
|
3, 4 |
741 |
|
NM_139211.4 |
NP_631957.1
|
2, 3 |
343 |
|
NM_139212.3 |
NP_631958.1
|
2, 3 |
616 |
|
XM_017008728.1 |
XP_016864217.1
|
2, 3 |
378 |
|
XM_017008729.1 |
XP_016864218.1
|
2, 3 |
260 |
|
XM_017008730.1 |
XP_016864219.1
|
2, 3 |
308 |
|
XM_017008731.1 |
XP_016864220.1
|
2, 3 |
343 |
|
XM_017008732.1 |
XP_016864221.1
|
3, 4 |
619 |
|
XM_017008733.1 |
XP_016864222.1
|
2, 3 |
183 |
|
XM_017008734.1 |
XP_016864223.1
|
2, 3 |
239 |
|
GenBank mRNA |
AB019573.1 |
- |
1, 2 |
53 |
|
AB059408.1 |
- |
2, 3 |
301 |
|
AB059409.1 |
- |
2, 3 |
184 |
|
AB059410.2 |
- |
2, 3 |
193 |
|
AF454763.1 |
- |
2, 3 |
206 |
|
AF492675.1 |
- |
2, 3 |
590 |
|
AF492676.1 |
- |
3, 4 |
322 |
|
AF492677.1 |
- |
3, 4 |
366 |
|
AF492680.1 |
- |
2, 3 |
342 |
|
AF492681.1 |
- |
2, 3 |
305 |
|
AK123318.1 |
- |
1 |
2413 |
|
AK289707.1 |
- |
2, 3 |
285 |
|
AW955211.1 |
- |
2, 3 |
309 |
|
BC014225.2 |
- |
1, 2 |
456 |
4335211 |
CB996541.1 |
- |
2, 3 |
379 |
|
EB386856.1 |
- |
2, 3 |
337 |
|
View Full Product Details