Gene Transcripts

Gene Symbol
GPHB5
Entrez Gene ID
122876
Gene Name
glycoprotein hormone beta 5
Gene Aliases
B5, GPB5, ZLUT1
Chromosome Location
Chr. 14: 63312835 - 63318879 on Build GRCh38
UniGene ID
Hs.375028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145171.3 NP_660154.3 2 185
GenBank mRNA AF403430.1 - 1 128
AF467770.1 - 1 128
BC069113.1 - 1 128 7216930
BC133702.1 - 1 128
BM709443.1 - 2 185

View Full Product Details