Gene Transcripts

Gene Symbol
TRMT61A
Entrez Gene ID
115708
Gene Name
tRNA methyltransferase 61A
Gene Aliases
C14orf172, GCD14, Gcd14p, TRM61, hTRM61
Chromosome Location
Chr. 14: 103529172 - 103537073 on Build GRCh38
UniGene ID
Hs.525610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152307.2 NP_689520.2 4 2186
GenBank mRNA AK097771.1 - 5 1687
BC007241.1 - 1 766
BC016033.2 - 3 1772

View Full Product Details