Gene Transcripts
- Gene Symbol
- RBX1
- Entrez Gene ID
- 9978
- Gene Name
- ring-box 1
- Gene Aliases
- BA554C12.1, RNF75, ROC1
- Chromosome Location
- Chr. 22: 40951347 - 40973015 on Build GRCh38
- UniGene ID
- Hs.474949
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014248.3 |
NP_055063.1
|
1, 2 |
130 |
|
GenBank mRNA |
AF085906.1 |
- |
1, 2 |
59 |
|
AF140598.1 |
- |
1, 2 |
84 |
|
AF142059.1 |
- |
1, 2 |
78 |
|
AK315722.1 |
- |
1, 2 |
89 |
|
AY099360.1 |
- |
1, 2 |
41 |
|
BC001466.2 |
- |
1, 2 |
83 |
3138751 |
BC017370.1 |
- |
1, 2 |
100 |
|
CR456560.1 |
- |
1, 2 |
96 |
|
DA353856.1 |
- |
1, 2 |
130 |
|
View Full Product Details