Gene Transcripts

Gene Symbol
Cebpd
Entrez Gene ID
25695
Gene Name
CCAAT/enhancer binding protein (C/EBP), delta
Gene Aliases
C/EBPd, CELF
Chromosome Location
Chr. 11: 89008008 - 89009146 on Build Rnor_6.0
UniGene ID
Rn.202620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013154.2 NP_037286.1 1 727
GenBank mRNA M65149.1 - 1 696

View Full Product Details