Gene Transcripts

Gene Symbol
Tmsb4x
Entrez Gene ID
81814
Gene Name
thymosin beta 4, X-linked
Gene Aliases
Chromosome Location
Chr. X: 28593405 - 28617267 on Build Rnor_6.0
UniGene ID
Rn.201192
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031136.1 NP_112398.1 3 191
GenBank mRNA BC058137.1 - 3 205
BC084720.1 - 3 180
BC097308.1 - 3 190
FQ211846.1 - 3 198
FQ211950.1 - 3 201
FQ213262.1 - 3 198
FQ214330.1 - 3 198
FQ217154.1 - 3 198
FQ217532.1 - 3 198
FQ217617.1 - 3 198
FQ217622.1 - 3 198
FQ220985.1 - 3 198
FQ221085.1 - 3 198
FQ221124.1 - 3 198
FQ221299.1 - 3 198
FQ221364.1 - 3 198
FQ221505.1 - 3 198
FQ221785.1 - 3 198
FQ221964.1 - 3 198
FQ222006.1 - 3 198
FQ222011.1 - 3 198
FQ222095.1 - 3 198
FQ222205.1 - 3 198
FQ222334.1 - 3 198
FQ222360.1 - 3 192
FQ222463.1 - 3 198
FQ222519.1 - 3 198
FQ222755.1 - 3 198
FQ223266.1 - 3 195
FQ223325.1 - 3 198
FQ228380.1 - 3 198
FQ228401.1 - 3 198
FQ228412.1 - 3 198
FQ228477.1 - 3 198
K01334.1 - 2 159
M26759.1 - 2 165
M34043.1 - 3 191

Gene Symbol
LOC103692338
Entrez Gene ID
103692338
Gene Name
putative thymosin beta-4-like protein 6
Gene Aliases
Chromosome Location
Chr. 5: 58535697 - 58536046 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763656.1 XP_008761878.1 1 220

View Full Product Details