Gene Transcripts

Gene Symbol
Gpr6
Entrez Gene ID
83683
Gene Name
G protein-coupled receptor 6
Gene Aliases
Chromosome Location
Chr. 20: 45813169 - 45815940 on Build Rnor_6.0
UniGene ID
Rn.32092
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031806.1 NP_113994.1 2 1377
GenBank mRNA AF064706.1 - 2 1377
U12006.1 - 2 1377

View Full Product Details