Gene Transcripts
- Gene Symbol
- Spag11a
- Entrez Gene ID
- 246305
- Gene Name
- sperm associated antigen 11A
- Gene Aliases
- Bin1b, Spag11, Spag11b
- Chromosome Location
- Chr. 16: 75666488 - 75668180 on Build Rnor_6.0
- UniGene ID
- Rn.81215
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF217088.1 |
- |
2 |
237 |
|
View Full Product Details