Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177934.3 | NP_808795.1 | 3 | 967 | |
XM_006242399.2 | XP_006242461.1 | 2 | 788 | ||
XM_008765749.1 | XP_008763971.1 | 3 | 844 | ||
GenBank mRNA | AY191521.1 | - | 3 | 972 |