Gene Transcripts

Gene Symbol
P2ry1
Entrez Gene ID
25265
Gene Name
purinergic receptor P2Y, G-protein coupled, 1
Gene Aliases
P2y, P2y1
Chromosome Location
Chr. 2: 151314818 - 151320312 on Build Rnor_6.0
UniGene ID
Rn.10217
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012800.1 NP_036932.1 1 1112
GenBank mRNA U22830.1 - 1 1112

View Full Product Details