Gene Transcripts
- Gene Symbol
- Htr4
- Entrez Gene ID
- 25324
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 4, G protein-coupled
- Gene Aliases
- Chromosome Location
- Chr. 18: 57637297 - 57820258 on Build Rnor_6.0
- UniGene ID
- Rn.10094
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012853.1 |
NP_036985.1
|
2, 3 |
171 |
|
XM_006254793.2 |
XP_006254855.1
|
3, 4 |
710 |
|
XM_008772089.1 |
XP_008770311.1
|
1, 2 |
1836 |
|
XM_008772090.1 |
XP_008770312.1
|
1, 2 |
1836 |
|
GenBank mRNA |
AJ011370.1 |
- |
3, 4 |
251 |
|
AM712909.1 |
- |
2, 3 |
151 |
|
AM712910.1 |
- |
2, 3 |
151 |
|
AM712911.1 |
- |
2, 3 |
151 |
|
U20906.1 |
- |
2, 3 |
171 |
|
U20907.1 |
- |
2, 3 |
171 |
|
View Full Product Details