Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164239.1 | NP_001157711.1 | 2 | 118 | |
NM_003587.4 | NP_003578.2 | 2 | 439 | ||
GenBank mRNA | AB001601.1 | - | 2 | 366 | |
AK302721.1 | - | 2 | 112 | ||
AK312264.1 | - | 2 | 378 | ||
BC002789.2 | - | 2 | 362 | ||
BC008825.2 | - | 2 | 390 | 4100320 | |
BC009392.2 | - | 2 | 390 | 4136069 | |
DB072495.1 | - | 2 | 440 |