Gene Transcripts
- Gene Symbol
- S100b
- Entrez Gene ID
- 25742
- Gene Name
- S100 calcium binding protein B
- Gene Aliases
- S100P
- Chromosome Location
- Chr. 20: 13130633 - 13142856 on Build Rnor_6.0
- UniGene ID
- Rn.8937
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013191.1 |
NP_037323.1
|
2, 3 |
266 |
|
XM_008772868.1 |
XP_008771090.1
|
2, 3 |
265 |
|
GenBank mRNA |
BC087026.1 |
- |
2, 3 |
204 |
|
FQ213381.1 |
- |
2, 3 |
208 |
|
FQ213687.1 |
- |
2, 3 |
258 |
|
FQ213774.1 |
- |
2, 3 |
260 |
|
FQ213840.1 |
- |
2, 3 |
216 |
|
FQ214218.1 |
- |
2, 3 |
210 |
|
FQ214292.1 |
- |
2, 3 |
239 |
|
FQ214554.1 |
- |
2, 3 |
208 |
|
M15705.1 |
- |
1, 2 |
133 |
|
M54919.1 |
- |
2, 3 |
266 |
|
X01090.1 |
- |
2, 3 |
266 |
|
View Full Product Details